Learn More
Bio-Serv™ Crawl Balls™ Enrichment Device, Certified
SDP
Catalog No. 14726538
Encompass
Change view
Click to view available options
Color:
Amber
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-726-538 Red
14-726-537 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-726-538 Supplier Bio-Serv™ Supplier No. K3329
Provides a comfortable hiding place for rodents
  • 4 in. (10.2cm) dia. with three 2.5 in. (6.4cm) openings
  • Made of high-temperature polycarbonate
  • Transparent for ease of visibility
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications
Autoclavable Autoclavable
Product Type Crawl Ball
Color Red
Material Polycarbonate
Diameter (English) Inside 4 in.
Diameter (Metric) Inside 10.16 cm