Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Fast-Trac Activity Wheel, Certified
SDP

Catalog No. 14726522
Encompass
Change view
Click to view available options
Color:
Amber
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-726-522 Amber
14-726-542 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-726-522 Supplier Bio-Serv™ Supplier No. K3250
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specialized activity wheel for use with the Mouse Igloo shelter (Mfr. Nos. K3328, K3570, and K3327)

  • Supports natural and instinctive running behavior
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)

Specifications

Autoclavable Autoclavable
Product Type Fast-Trac Wheel Activity
Color Amber
Material Polycarbonate
For Use With (Application) Supports natural and instinctive running behavior
For Use With (Equipment) Mouse igloo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.