Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Mouse Diet, High Fat Paste
SDP

For induction and/or maintenance of obese models

$373.86 - $389.26

Specifications

Calories 5.51
Components Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
For Use With (Application) Induction and or maintenance of obese models
Particle Shape Paste
Quantity 10,000 g
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Sterility Price Quantity & Availability  
14-726-604
SDP
Encompass
View Documents
Bio-Serv™
F1850
Mouse Diet Non-sterile
Each for $373.86
Only null left
Add to Cart
 
14-726-605
SDP
Encompass
View Documents
Bio-Serv™
S1850
Mouse Diet Sterile
Each for $389.26
Only null left
Add to Cart
 
Description

Description

  • Nutritionally complete purified diet formula successfully used in diabetes and obesity studies
  • 60% kcals from fat
  • Nutritionally assayed
  • Ingredients: Lard, Sucrose, Casein, Maltodextrin, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride

Specifications

Specifications

5.51
Induction and or maintenance of obese models
10,000 g
Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
Paste
Mouse
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.