Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio-Serv™ Mouse Diet, High Fat, Soft Pellets

For induction and/or maintenance of obese models
$198.40 - $213.72
Specifications
Calories | 5.49 |
---|---|
Components | Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride |
Format | Pellet |
For Use With (Application) | Nutritionally complete purified diet formula successfully used in diabetes and obesity studies |
Particle Shape | Soft Pellet |
Catalog Number | Mfr. No. | Product Type | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Sterility | Price | Quantity & Availability | ||||
14-726-510
![]() ![]() |
Bio-Serv™
F3282 |
Mouse Diet | Non-sterile |
Each for $198.40
|
|
||||
14-726-603
![]() ![]() |
Bio-Serv™
S3282 |
Mouse Diet | Sterile |
Each for $213.72
|
|
||||
Description
- Nutritionally complete purified diet formula successfully used in diabetes and obesity studies
- 60% kcals from fat
- Soft pellets are 0.5 in. dia. (1.3cm) and vary in length
- Nutritionally assayed
- Ingredients: Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
Specifications
5.49 | |
Pellet | |
Soft Pellet | |
Mouse | |
1.27 cm |
Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride | |
Nutritionally complete purified diet formula successfully used in diabetes and obesity studies | |
5000 g | |
0.5 in. |
Safety and Handling
ShelfLife : 6 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title