Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Mouse Igloo™ Rodent Enrichment Device, Certified
SDP

Catalog No. 14726502
Encompass
Change view
Click to view available options
Color:
Amber
Blue
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-726-502 Amber
14-726-552 Red
14-726-551 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-726-502 Supplier Bio-Serv™ Supplier No. K3328
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Provides shelter and a secluded place to build a nest

Provides shelter and a secluded place to build a nest

  • Transparent material provides ease of visibility
  • Great addition to breeding programs
  • Fits most shoebox cages
  • Stacks easily for convenient storage
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)

Specifications

Autoclavable Autoclavable
Product Type Mouse Igloo
Color Amber
Material Polycarbonate
For Use With (Application) Provide shelter and a secluded place to build a nest, great addition to breeding programs
Height (English) 2.25 in.
Height (Metric) 5.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.