Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ PRANG™, Oral Rehydrator Powder
SDP

Catalog No. 14726437
Encompass
Change view
Click to view available options
Quantity:
100 g
1000 g
Flavor:
Apple
Banana
Cherry
Grape
Orange
Strawberry
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flavor
14-726-437 1000 g Apple
14-726-431 100 g Apple
14-726-434 100 g Grape
14-726-435 100 g Orange
14-726-439 1000 g Cherry
14-726-438 1000 g Banana
14-726-440 1000 g Grape
14-726-441 1000 g Orange
14-726-442 1000 g Strawberry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-726-437 Supplier Bio-Serv™ Supplier No. F2351A
Add to Cart
Add to Cart

Excellent for crashing, recovering and debilitation conditions

  • Flavored oral rehydration with electrolytes and Vitamin C
  • Balanced mineral and glucose levels for quick energy
  • Provides the benefit of electrolyte drinks with less sugar
  • Powder form, just add water
  • Nutritionally assayed
  • Certified (contaminant screened)

Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye

Specifications

Product Type Oral Rehydrator
Calories 19.88
Components Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
Flavor Apple
Format Powder
For Use With (Application) Crashing, recovering, debilitating
Particle Shape Powder
Quantity 1000 g
Storage Requirements Cool dry place
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.