Learn More
Bio-Serv™ PRANG™, Oral Rehydrator Powder

Description
- Flavored oral rehydration with electrolytes and Vitamin C
- Balanced mineral and glucose levels for quick energy
- Provides the benefit of electrolyte drinks with less sugar
- Powder form, just add water
- Nutritionally assayed
- Certified (contaminant screened)
Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
Specifications
Specifications
| Product Type | Oral Rehydrator |
| Calories | 19.88 |
| Components | Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye |
| Flavor | Grape |
| Format | Powder |
| For Use With (Application) | Crashing, recovering, debilitating |
| Particle Shape | Powder |
| Quantity | 1000 g |
| Storage Requirements | Cool dry place |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.