Learn More
Agilent BioTek Bio-Stack Twister II Accessories
Catalog No. BT48634 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Bio-Stack Twister II
Catalog No. For Use With (Equipment)
BT48634 Bio-Stack Twister II
1 options
Catalog No. BT48634 Supplier Agilent BioTek Supplier No. 48634
Expand the capacity and capability of the Twister II
  • The KC4 integration kit is for use with the PowerWave Microplate Spectrophotometer, Clarity Luminescence Reader, and the Synergy HT Multi-detection Reader
  • The GEN5 Kit supports the PowerWave Microplate Spectrophotometer as well as the Synergy HT and Synergy 2 Multi-detection Readers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Bio-Stack Twister II