Learn More
Leica Biosystems Bio Wraps, 2X3in, locate small biopsy specimens following paraffin infiltration, 500/bx
Supplier:  Leica Biosystems 3801090
Encompass
Bio-Wraps are sections of tissue paper cut to a convenient, ready-to-use 2 in. x 3 in. size. They are colored blue to help locate small biopsy specimens following paraffin infiltration. They will not bleed into specimens. Bio-Wraps are available in convenient dispenser packs.
Catalog No. NC9973283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.