Learn More
Bio X Cell RecombiMAb Human IgG4 (S228P) Isotype Control Antibody, Anti-Hen Egg Lysozyme, 25 mg (Bio X Cell CP147)
Supplier: Bio X Cell CP147R025MG
RecombiMAb human IgG4 (S228P) isotype control antibody reactive with hen egg lysozyme, produced recombinantly in CHO cells and supplied as a purified antibody for use as an isotype control in antibody-based assays. The S228P mutation prevents IgG4 Fab arm exchange; formulation is PBS pH 7.0, purity ≥95%, supplied as a 25 mg vial; store at 4°C and do not freeze.
- Recombinant human IgG4 with S228P mutation.
- Reacts with hen egg lysozyme; minimal cross-reactivity to human samples.
- Produced in CHO cells in an animal-free facility.
- Purity ≥95% as determined by SDS-PAGE.
- Formulated in PBS, pH 7.0, no stabilizers or preservatives.
- Supplied as a 25 mg vial; store at 4°C, do not freeze.
- Intended for use as an isotype control in antibody assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.