Learn More
Alpco Ltd Bioactive Leptin ELISA
SDP
Supplier:  Alpco Ltd 22BLEPHUE01
Encompass_Preferred
The Bioactive Leptin ELISA is for the quantitative measurement of functional, receptor binding Leptin levels in serum or plasma. 
  • Analytical Sensitivity: < 0.01 ng/mL
  • Dynamic Range: 0.05 - 6 ng/mL
  • Sample Size: 15 µL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2318943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.