Learn More
Thermo Scientific™ BioBasic™ 4 5μm Particle Size Drop-In Guard Cartridges
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ BioBasic 4 5μm Particle Size Drop-In Guard Cartridges offer convenience, economy and effective protection for extending column lifetime.
- The 10mm design offers maximum protection with minimal increase in retention Fit Thermo Scientific Uniguard™ direct connection and stand alone holders
- Based on 300Å silica for outstanding biomolecule performance
- Lower carbon load for optimal retention of larger peptides and proteins
- Outstanding reproducibility, efficiency and column lifetime
- Ideal for LC/MS separations
- Available in packs of 4
Requires:
Thermo Scientific Uniguard™ direct connection or stand-alone holder
Specifications
Specifications
| Carbon Load | 4% |
| pH | 2 to 10 |
| Particle Size | 5 μm |
| Length (Metric) | 10 mm |
| Pore Size | 300Å |
| Diameter (Metric) | 4.6 mm |
| For Use With (Equipment) | Uniguard Direct-connection and Stand Alone Holder |
| Endcapped | Yes |
| USP Type | L26 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.