Learn More
Biocare Medical, LLC p63 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM163C
Encompass_Preferred
p53 homologue p63 encodes for different isotypes able to either transactivate p53 reporter genes (TAp63) or act as p53-dominant-negatives. p63 is detected in prostatic basal cells in normal prostate; however, it is negative in malignant tumors of the prostate gland. Thus p63 antibody may be a valuable tool in the differential diagnosis of benign and malignant tumors of prostate gland and can be used in a panel of antibodies such as HMW CK [34ßE12], PSA and PSAP. p63 may play a significant role in prostate development by maintaining a prostate stem cell population. Striated muscle staining may be observed with p63.
Catalog No. NC0758257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.