Learn More
Vernier™ BioChamber 2000
SDPSCI_ED
Catalog No. S16083ND Shop All Vernier Software Products
Change view
Click to view available options
Description:
2000 mL Capacity
Catalog No. Description
S16083ND 2000 mL Capacity
1 options
Catalog No. S16083ND Supplier Vernier™ Supplier No. BC2000
Add to Cart
Add to Cart
Includes
BioChamber 2000
The BioChamber 2000 is a 2000 mL plastic chamber that can be used with a Gas Sensor and an Gas Sensor to monitor gaseous carbon dioxide and oxygen levels simultaneously in a closed system. The BioChamber 2000 is particularly useful when monitoring respiration and photosynthesis. The large top opening provides easy access to insert, adjust, or remove the sample.
  • Particularly useful when monitoring respiration and photosynthesis
  • Large top opening provides easy access to insert, adjust, or remove the sample
  • Includes two sensor ports
  • If you choose not to use both ports, a size 6 rubber stopper fits snugly in the grommet
  • Stopper does not guarantee a water tight seal
Warranty and Services
None
Specifications
Description 2000 mL Capacity
Interface Two sensor ports that fit a size 6 rubber stopper
Material Plastic
Warranty 5 Year Limited
For Use With (Application) Monitoring respiration and photosynthesis
For Use With (Equipment) CO2 Gas Sensor, Ethanol Sensor, O2 Gas Sensor
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov