Learn More
Vernier™ BioChamber 250
SDPSCI_ED
Catalog No. S16084ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
CO2 Gas Sensor, O2 Gas Sensor
Catalog No. For Use With (Equipment)
S16084ND CO2 Gas Sensor, O2 Gas Sensor
1 options
Catalog No. S16084ND Supplier Vernier™ Supplier No. BC250
Add to Cart
Add to Cart
Includes
BioChamber 250
The BioChamber 250 is a 250 mL Nalgene bottle that can be used with a Gas Sensor and an Gas Sensor to monitor gaseous carbon dioxide and oxygen levels simultaneously in a closed system. The BioChamber is useful in experiments such as photosynthesis and cell respiration.
  • Nalgene plastic bottle
  •  Useful in experiments such as photosynthesis and cell respiration
  • With the bottle on its side, place the CO2 Gas Sensor into the neck of the BioChamber and the O2 Gas Sensor into the rubber grommet as shown
  • This design keeps the O2 Gas Sensor in the desired upright position while placing the CO2 Gas Sensor in an ideal position for gas level monitoring
  • A size 6 rubber stopper fits into the rubber grommet
  • Stopper does not guarantee a water tight seal
  • Replaces the CO2 to O2 Tee
Warranty and Services
None
Specifications
Description 250 mL Capacity
Interface One sensor port that fit a size 6 rubber stopper
Material Plastic
Warranty 5 Year Limited
For Use With (Application) Monitoring respiration and photosynthesis
For Use With (Equipment) CO2 Gas Sensor, O2 Gas Sensor
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov