Learn More
Alkali Scientific BioClave B4000-28-120 Research
SDP
Supplier:  Alkali Scientific RS7524
Encompass_Preferred
BIOCLAVERESEARCH AUTOCLAVES STERILIZER Fully automatic just press start Rapid complete sterilization Extremely compact Built in water tank does not require an external water supply Despite the space-saving exterior dimensions of the Bioclave B4000-28-120 the stainless steel chambers are surprisingly large accommodating a variety of liquids media instruments glassware plasticware and other common research laboratory items Available in 8L 18L and 28L chamber volumes these benchtop models include a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI With a large digital LCD display and fully automatic operation all segments of the cycle (fill sterilize exhaust and dry) commence and run to completion by simply pressing the start button An optional thermal mini-printer is also available for providing a record of sterilization parameters and a USB drive can be used to record settings and transfer them to a PC The Bio
Catalog No. 50-302-9630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.