Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell BioClean-C Chemotherapy Protective Sleeve Covers

Catalog No. 19668258 Shop All Ansell Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-668-258 Non-sterile
19-668-257 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-668-258 Supplier Ansell Supplier No. BCSC
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioClean-C sleeve covers provide protection against a wide range of cytotoxic drugs. Manufactured from lightweight and low-linting CleanTough™ material.

  • Elasticated for secure fit
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Certifications and testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
    • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
    • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
    • Tested against standard EN 14325 protective clothing against chemicals
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
    • Tested against standards: EN 530, Abrasion resistance; EN 6530,
    • Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environment Sleeve
Size Standard
Construction Ultrasonically bonded seams covered with protective tape
Material Microporous Fabric
Color Blue
Sterility Non-sterile
Closure Type Elastic Ends
Seam Type Ultrasonically Bonded with Protective Tape
Cuff Style Elastic
Model BCSC
For Use With (Application) Cleanroom, Clinical, Medical, Laboratory, Pharmaceutical
For Use With (Equipment) BioClean-C™ Sterile Chemotherapy Protective Aprons
Certifications/Compliance ASTM D6978-05, ASTM F739-12
Certifications/Compliance (Europe) EN ISO 13034:2005 + A1:2009, EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Cleanroom Class Class 10 (ISO 4)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.