Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ BioClean™ Clearview Autoclavable Panoramic Goggles Replacement Strap

Catalog No. 19668275 Shop All Ansell Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-668-275 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-668-275 Supplier Ansell™ Supplier No. BCAPSS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ansell™ BioClean™ Clearview Autoclavable Panoramic Goggles Replacement Strap

  • Material: Silicone
  • Material of hooks: Polypropylene
  • Includes adjuster hooks and strap clips
  • Packaging: 25 pieces per sealed PE bag; two bags per inner box; 10 boxes per carton
  • Shelf life: 5 years
  • Storage: Keep away from direct sunlight, store in a dry place, keep in original packaging, and keep away from ozone sources
TRUSTED_SUSTAINABILITY

Specifications

Description Replacement Strap for BCAP Autoclavable Panoramic Goggles
Color Clear
Product Type Replacement part
Quantity 1 Ea.
Disposable Reusable
Frame Material Silicone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.