Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell BioClean-D™ BDLC Sterile Lab Coats

Catalog No. 17010332 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-332 Medium
17-010-333 Small
17-010-331 Large
17-010-334 X-Large
17-010-335 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-010-332 Supplier Ansell Supplier No. SBDLCM
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured from antistatic lightweight CleanTough material and features press stud fastenings, open cuffs, three pockets, and a rear vent.

  • Lightweight CleanTough material
  • Press stud fastening
  • Open cuffs and rear vent
  • Three deep pockets
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polyethylene, Non-Woven Polypropylene Laminate
Color White
Closure Type Snaps
Cuff Style Open
Disposable Limited-use
Sterility Sterile
Resistance Abrasion, Tears, Liquids, Chemicals
Style Knee-length
No. of Pockets 3
Latex Free Yes
Model S-BDLC
Seam Type Bound
Certifications/Compliance ASTM F739, ISO 11137-1
Certifications/Compliance (Europe) EN 1149-5, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325
For Use With (Application) Laboratory, Research, Veterninary Services
Autoclavable Not Autoclavable
Construction Bound Seams, Single Needle Stitching
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.