Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ BioClean-D Drop-Down Garment with Hood

Catalog No. 19668239 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-668-239 Medium
19-668-244 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-668-239 Supplier Ansell™ Supplier No. BDSHM
This item is not returnable. View return policy

Ansell™ BioClean-D Drop-Down Garment with Hood features quick release press stud tabs for aseptic donning

  • Elasticated back, hood, cuffs and ankles
  • Low-linting CleanTough material
  • Foot-loop to aid smooth closure of zip
  • Thumb loops to ensure a secure hold
  • Silicone free
  • Certifications and testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
    • Tested against standards: EN 1149-5:2008, protective clothing - electrostatic properties; EN 530, Abrasion resistance; EN 7854, Flex cracking resistance; EN 6530, Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Material CleanTough™ Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static, Abrasion, Tears
Collar Type Hooded
Back Style Elastic
No. of Pockets None
Size (English) Chest 36 to 39 in.
Size (Metric) Chest 92 to 100 cm
Sleeve Style Elastic
For Use With (Application) Controlled Environments
Height (English) 5 ft. 6 in. to 5 ft. 9 in.
Quantity 20/Cs.
Style Drop-down
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.