Learn More
Ansell BioClean-D Long-Length Non-Sterile Hood
BIOPHARMA
Catalog No. 19500863 Shop All Ansell Products
Change view
Click to view available options
Material:
Spunbond Polypropylene
Catalog No. Material
19-500-863 Spunbond Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 19-500-863 Supplier Ansell Supplier No. BDHDL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Non-sterile hood features an extra-long yoke for maximum coverage.
  • Lightweight low-linting CleanTough material
  • ESD properties
  • PPE Cat 3 Type PB [6]
  • Silicone free
  • Certifications and Testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425
    • Module D certificationsConforms to Category 3 - Personal Protective Equipment Regulation(EU)2016/425
    • Tested against standard EN ISO 13034:2005+A1:2009 protective clothing against liquid chemicals (Type PB[6])
    • Tested against standard EN ISO 14325 protective clothing against chemicals
    • Tested against standards: EN 1149-5:2008, protective clothing -electrostatic properties; EN 530, Abrasion resistance; EN 7854, Flexcracking resistance; EN 6530, Resistance to penetration by liquids; EN13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN863, Puncture resistance; and EN 13935-2, Seam strength.
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hood
Size Universal
Material Spunbond Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Design Type Three-piece
Model BDHD
Seam Type Bound, Single Needle Stitching
Sterility Non-sterile
Quantity 120/Pk.
Hood Type Three-piece, Elastic face-opening
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.