Learn More
Ansell BioClean-D™ Sterile Hood
BIOPHARMA
Catalog No. 19500864 Shop All Ansell Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
19-500-864 Polypropylene
1 options
This item is not returnable. View return policy
N/A
Catalog No. 19-500-864 Supplier Ansell Supplier No. SBDHDL
Add to Cart
Add to Cart
This item is not returnable. View return policy
N/A
Long length cleanroom hood features an extra-long yoke for maximum coverage.
  • Extended length: More complete protective coverage
  • Low-linting lightweight material: Fewer contamination risks
  • ESD properties: Reduced risk of electrostatic interference or damage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sterile Hood
Size Universal
Material Polypropylene
Color White
Closure Type Pull-over
Disposable Single-use
Latex Free No
Model S-BDHD
Seam Type Bound
Size (English) Universal
Size (Metric) Universal
Sterility Sterile
Construction Low-linting, Lightweight Material
For Use With (Application) Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical
Resistance Electrostatic Discharge
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
N/A