Learn More
Ansell BioClean-D™ Non-Sterile Cleanroom Overboots
Catalog No. 19501004 Shop All Ansell Products
Change view
Click to view available options
Style:
Long Length
Standard Length
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-501-004 Long Length
19-668-318 Standard Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
N/A
Catalog No. 19-501-004 Supplier Ansell Supplier No. BDOBL
Add to Cart
Add to Cart
This item is not returnable. View return policy
N/A
Low-linting antistatic overboots offer added protection with a firm, elasticated fit. Available in regular or long length sizes.
  • Lightweight low-linting CleanTough™ fabric: Lower contamination risks
  • Elasticated opening: A secure fit, for enhanced personal protection
  • Tie fastenings and slip-resistant sole: Secure fit and safe use
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Ties
Sole Type Slip-resistant
Construction Bound Seams, Single Needle Stitching
Closure Color White
Seam Type Bound
Style Long Length
Thread Color White
For Use With (Application) Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical
Certifications/Compliance ISO 9001:2015, ASTM F739-12
Certifications/Compliance (Europe) (EU)2016/425, EN ISO 13034:2005 +A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Packaging Quantity 30 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; five outer bags per lined carton (150 pieces)
Height (Metric) 500 mm
Resistance Dry Particulates, Liquid, Chemical-resistant, Chemical-resistant
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
N/A