Learn More
Ansell BioClean-D™ Sterile Disposable Overboots
Catalog No. p-9023019 Shop All Ansell Products
Change view
Click to view available options
Style:
Long Length
Standard Length
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-668-317 Standard Length
19-501-005 Long Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
N/A
Catalog No. 19-668-317 Supplier Ansell Supplier No. SBDOB
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
N/A
Lightweight sterile overboots with an elasticated opening provides secure, fitted personal protection. Available in regular or long length sizes.
  • Lightweight low-linting CleanTough™ fabric: Fewer contamination risks
  • Elasticated opening: A more secure, more comfortable fit
  • Slip-resistant sole and easy tie fastenings: Secure fit, for safer use
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material CleanTough™
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Ties
Sole Type Slip-resistant
Construction Bound Seams, Single Needle Stitching
Closure Color White
Seam Type Bound
Style Standard Length
Thread Color White
For Use With (Application) Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical
Certifications/Compliance PPE Cat 3 Type PB [6]; ISO Class 4 and EU GMP Grade A; Anti-static to EN1149-5* and low particulating
Certifications/Compliance (Europe) PPE Cat 3 Type PB [6]; ISO Class 4 and EU GMP Grade A; EN1149-5
Packaging Quantity One pair per sealed inner PE bag; 15 inner bags per sealed outer PE bag; five outer bags per lined carton (75 pairs)
Resistance Liquid, Dry Particulates, Splash
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
N/A