Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell BioClean™ S-BGHF Sterile Half-Finger Nylon Glove Liners

Catalog No. 19500955 Shop All Ansell Products
Change view
Click to view available options
:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-500-955 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-500-955 Supplier Ansell Supplier No. SBGHF
This item is not returnable. View return policy

Glove liners are designed specifically to provide a comfortable barrier between the skin and the outer glove. Suitable for use in Class 10/ISO 4 Cleanrooms.

  • Half-finger style for maximum tactility
  • Provides a soft barrier between the skin and glove
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EU GMP Grade A
Cleanroom Class Class 10 (ISO 4)
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nylon
Size Universal
Sterility Sterile
Finger Style Half
For Use With (Application) Life Sciences
Glove Cuff Style Beaded
Series BGHF
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.