Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Endothelial Cell Growth Environment Kit

Catalog No. 08774149AA Shop All Corning Products
Change view
Click to view available options
Type:
Endothelial Cell Culture Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
08-774-149AA Endothelial Cell Culture Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-149AA Supplier Corning™ Supplier No. 355054

Includes

Complete Environment Kit: 500mL BD Endothelial Cell Culture Medium (MCDB 131 medium containing hydrocortisone, heparin and 2% fetal bovine serum; 5μg lyophilized Mouse Epidermal Growth Factor (EGF), receptor grade; 100mg lyophilized Bovine Endothelial Cell Growth Supplement (ECGS); 50mg lyophilized Soybean Trypsin Inhibitor; Five BD BioCoat Type I Collagen 75cm2 Flasks

Increases endothelial cell yield from a variety of sources

Ideal for human umbilical vein (HUVEC), pulmonary artery (HPAEC), and aorta (HAEC), as well as fetal bovine heart (FBHEC) cells. Interactions between components of the extracellular matrix and growth factors play a crucial role in determining the adaptation and responses of mammalian cells to their micro environments both in vivo and in vitro.

  • Cells can be subcultured according to routine procedures, seeded into fermentation systems, or used in a variety of bioassays
  • Integrate specially formulated low-serum or serum-free medium, culture supplements and growth factors
  • Each environment lot is tested in a bioassay and all components are tested for bacteria and fungi

These products are for research use only and are not intended for diagnostic purposes.

Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology

Order Info

Media and cellware are also available separately.

TRUSTED_SUSTAINABILITY

Specifications

Type Endothelial Cell Culture Medium
For Use With (Application) Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Storage Requirements 2° to 8°C
No. per Pack 1/Pk.
No. per Case 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.