Learn More
Description
Specifications
Specifications
| Capacity (English) | 25.3 oz. |
| Closure Material | Phenolic |
| Capacity (Metric) | 750 mL |
| Material | Polystyrene |
| For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
| Neck Style | Canted |
| No. per Case | 5/cs. |
| Shape | Rectangular |
| Surface Coating | Poly-D-Lysine |
| Sterility | Sterile |
| Show More |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
