Learn More
BIOSMITH BIOCOOLER ALUMINUM BLOCK
Supplier: BIOSMITH 81001
96 Well Aluminum Block Includes non-skid feet. 11.43cm x 7.62cm x 2.16cm These aluminum blocks are very useful for holding lightweight 96 well "V" bottom trays and 0.2ml tubes during sample preparation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.