Learn More
Biotium Biocytin (E-Biotinoyl-L-Lysine)
Supplier:  Biotium 90055
Encompass_Preferred
Biocytin (e-Biotinoyl-L-lysine) is a very useful cellular tracer that can be introduced into cells by microinjection.Properties: White solid soluble in water or DMSO; Store at 4°C; C16H28N4O4S; MW: 372.48; [576-19-2]
Catalog No. 50-196-4810
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.