Learn More
Research Products International Corp BioFlsk Erlnmyr Flask Baff Bot
SDP
Supplier:  Research Products International Corp 400854
Encompass_Preferred
BioFlast Cell Culture Erlenmeyer Flasks are made from a variety of disposable and autoclavable plastic. Vented screw caps (polypropylene) feature 0.2mm membranes to increase air transmission and enhance cell growth. Polycarbonated flasks and Jumbo Flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiently even at lower shaking speeds.
Catalog No. 50-238-8429
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.