Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Biofreeze™ Pain-Relieving Gel

Catalog No. 19090848 Shop All Medique Products
Change view
Click to view available options
Format:
Packet
Pump
Tube
Volume (English):
0.176 oz.
4 oz.
16 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Volume (English)
19-090-848 Pump 16 oz.
19-090-847 Tube 4 oz.
19-090-846 Packet 0.176 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-848 Supplier Medique Supplier No. 31116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cold therapy pain reliever.

  • For arthritis, back pain, sore muscles and joints

Specifications

Product Type Pain Relieving Gel
For Use With (Application) Cold therapy pain reliever for arthritis, back pain, sore muscles and joints
Format Pump
Quantity 1 Ea.
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.