Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Biohazard Disposable Bags
SDPSCI_ED

Catalog No. S139252 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
18 in.
22 in.
24 in.
24.5 in.
31.5 in.
35 in.
42 in.
Width (English):
12 in.
16 in.
18 in.
24 in.
25 in.
42 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Width (English)
S139252 35 in. 25 in.
S139248 18 in. 12 in.
S139249 24 in. 12 in.
S139250 22 in. 18 in.
S139253 31.5 in. 24 in.
S139254 42 in. 42 in.
S139251 24.5 in. 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Prop 65
Catalog No. S139252 Supplier United Scientific™ Supplier No. UNU40104
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prop 65

Biohazard bags provide a safe method of disposal for biohazard waste.

  • Bags have a clearly printed biohazard symbol and pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
  • Steam autoclavable for sterilization of waste before disposal.
  • Polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
  • High temperature steam autoclavable to 135°C (275°F) Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests 55 microns (2.2 mil) thickness

Specifications

Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Equipment) Biohazard Materials
Material Polypropylene
Sterility Non-sterile
Length (English) 35 in.
Width (English) 25 in.
Product Type Benchtop Biohazard Disposal Bag
Capacity (Metric) 9 L
Width (Metric) 635 mm
Show More Show Less

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.