Learn More
Description
- Bags have a clearly printed biohazard symbol and pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
- Steam autoclavable for sterilization of waste before disposal.
- Polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
- High temperature steam autoclavable to 135°C (275°F) Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests 55 microns (2.2 mil) thickness
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Red |
| Disposable | Single-use |
| For Use With (Equipment) | Biohazard Materials |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Length (English) | 31.5 in. |
| Width (English) | 24 in. |
| Product Type | Benchtop Biohazard Disposal Bag |
| Capacity (Metric) | 9 L |
| Show More |
Warning: Cancer and reproductive harm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

