Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Biohazard Disposal Bags
GREENER_CHOICE

Catalog No. 03448273
Change view
Click to view available options
Product Type:
Bag Holder
Benchtop Biohazard Disposal Bag
Capacity (Metric):
2 L
9 L
19 L
40 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Capacity (Metric)
03-448-273 Benchtop Biohazard Disposal Bag 19 L
03-448-271 Benchtop Biohazard Disposal Bag 2 L
03-448-272 Benchtop Biohazard Disposal Bag 9 L
03-448-274 Benchtop Biohazard Disposal Bag 40 L
03-448-270 Bag Holder 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-448-273 Supplier Heathrow Scientific™ Supplier No. HS10322
Add to Cart
Add to Cart

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag

Specifications

Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Application) Biohazard Disposal
For Use With (Equipment) Autoclave
Material Polypropylene
Sterility Non-sterile
No. per Case 500 Pk.
Outer Packaging Box
Thickness (English) 63 mil
Dimensions (L x W) 19 x 23 in.
Dimensions (L x W x H) 19 x 23 x 0.06 in.
Length (English) 19 in.
Width (English) 23 in.
Height (English) 0.06 in.
Height (Metric) 0.15 cm
Weight (English) 0.04 lb.
Weight (Metric) 0.02 kg
Product Type Benchtop Biohazard Disposal Bag
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Length (Metric) 48.3 cm
Width (Metric) 58.4 cm
Thickness (Metric) 1.6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.