Learn More
Bel-Art™ Biohazard Disposal Bags with or without Warning Label
Shop All SP Bel-Art & SP Wilmad-LabGlass ProductsDescription
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
- These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Specifications
Specifications
| Capacity (Metric) | 40 L |
| Certifications/Compliance | ASTM D1709-98 |
| Closure Type | Zip Tie |
| Design | Biohazard Symbol w/ Sterilization Patch |
| Capacity (English) | 10.5 gal. |
| Color | Clear |
| Dimensions (L x W) | 36 x 24 in. (91 x 61 cm) |
| Disposable | Single-use |
| Length (Metric) | 91 cm |
| Sterility | Non-sterile |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.