Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BIOHAZARD Label

Catalog No. 19129317
Click to view available options
Dimensions (LxW):
2.875 x 2 in.
5 x 3.5 in.

Self-sticking polyester BIOHAZARD Label is used to identify the actual or potential presence of a biologically hazardous material.

  • Good resistance to most common chemicals, oils and water.
  • Service temperature of -40°F to +122°F (-40°C to +50°C).
  • Black on Orange.
  • Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling.
  • Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation.

Specifications

Autoclavable Not Autoclavable
Adhesive Type Acrylic
Color Orange
Material Polyester
Printable Sides Front
Product Type Labels
Sterility Non-sterile
For Use With (Application) Hazardous Material Identification
Includes Pack of 25 Labels
Languages English
Quantity 25 Pk.
Shape Rectangle
Size (English) 2.875 x 2 in.
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 11 mil
Thickness (Metric) 0.279 mm
Dimensions (LxW) 2.875 x 2 in.
Width (English) 2 in.
Width (Metric) 50.80 mm
Height (English) 2.875 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.