Learn More
Market Lab MarketLab X-Large Biohazard Shield with Wrap Design, 1 each
Supplier:  Market Lab 16493
Encompass_Preferred
Wrapped design with side braces create strong platform for placement of items on top of shield. Protects and contains work area, including benchtop surfaces. Keeps everything contained in smaller space. Rubberized feet prevent sliding. Made of clear 3/16" acrylic.
Catalog No. NC1757462