Learn More
New Pig Biohazard Spill Kit
SDP
Catalog No. 18389149 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Biohazard Spill Kit
Catalog No.
18-389-149 Type: Biohazard Spill Kit
1 options
This item is not returnable. View return policy
Catalog No. 18-389-149 Supplier New Pig Supplier No. PLS1850
Add to Cart
Add to Cart
This item is not returnable. View return policy
Be prepared for biohazard spill response to absorb blood, urine, vomit and feces with this multi-use kit
  • Keep this portable, multi-use kit on hand to help reduce the risks while cleaning up blood, urine or other biofluids
  • Includes PPE, PIG Absorbent Pads, PIG Absorb-&-Lock Bio-Fluids Loose Absorbent, germicidal wipes and other supplies for responder and scene cleanup Ideal for first responders, hospitals, schools, nursing homes and industrial facilities
  • Packaged in a UN-rated container that can be used for shipping regulated wastes for disposal
  • Carrying handle makes kit easy to transport during emergency response