Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Biohazard Wipe Dispenser
SDP

Catalog No. 22000597
Change view
Click to view available options
Mounting:
Magnetic Mount
Tape
Wall or Surface Mount
Dimensions (L x W):
3 x 3 in.
4 x 4 in.
No. of Compartments:
1
2
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Dimensions (L x W) No. of Compartments
22-000-597 Magnetic Mount 3 x 3 in. 2
22-000-592 Wall or Surface Mount 3 x 3 in. 1
22-000-593 Tape 3 x 3 in. 1
22-000-594 Magnetic Mount 3 x 3 in. 1
22-000-595 Wall or Surface Mount 3 x 3 in. 2
22-000-596 Tape 3 x 3 in. 2
22-000-598 Wall or Surface Mount 4 x 4 in. 1
22-000-599 Tape 4 x 4 in. 1
22-000-600 Magnetic Mount 4 x 4 in. 1
22-000-601 Wall or Surface Mount 4 x 4 in. 2
22-000-602 Tape 4 x 4 in. 2
22-000-603 Magnetic Mount 4 x 4 in. 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 22-000-597 Supplier Poltex Supplier No. BHZDWIP32M
Add to Cart
Add to Cart

Made from super-durable and cleanable PETG, this Biohazard Wipe Dispenser holds a stack of biohazard wipes.

  • Choose from 1 or 2 column capacity
  • Choose your mounting method: Wall mount (screws/anchors included), magnet (powerful magnets included), VHB tape (very high bond tape included)

Specifications

Dimensions (L x W) 3 x 3 in.
Material PETG
Mounting Magnetic Mount
No. of Compartments 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.