Learn More
Poltex Biohazard Wipe Dispenser
Description
- Choose from 1 or 2 column capacity
- Choose your mounting method: Wall mount (screws/anchors included), magnet (powerful magnets included), VHB tape (very high bond tape included)
Specifications
Specifications
| Dimensions (L x W) | 4 x 4 in. |
| Material | PETG |
| Mounting | Wall or Surface Mount |
| No. of Compartments | 2 |