Learn More
Medique Biolife QR WoundSeal™ Powder
Catalog No. 19090600 Shop All Medique Products
Change view
Click to view available options
Quantity:
1 Application
2 Applications
For Use With (Application):
Easy-to-reach Wounds
Hard-to-reach Wounds
Large Wounds
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
19-090-600 1 Application Large Wounds
19-090-583 2 Applications Hard-to-reach Wounds
19-090-582 2 Applications Easy-to-reach Wounds
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-600 Supplier Medique Supplier No. 2330
Add to Cart
Add to Cart
Topical powder to stop wound bleeding immediately.
  • For use on easy-to-reach or hard-to-reach wounds
  • Stops bleeding in seconds
  • Forms protective seal over wound
  • Used by hospitals, sports teams, EMTs, US Military
Specifications
For Use With (Application) Large Wounds
Product Type Bleeding Control Powder
Quantity 1 Application