Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Biomass Detection Cuvette BTCUV, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S27251 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Biomass Detection Cuvette BTCUV, Sold by AquaPhoenix Scientific
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S27251 Biomass Detection Cuvette BTCUV, Sold by AquaPhoenix Scientific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27251 Supplier 3M™ Supplier No. BTCUV

missing translation for 'validMainframeMsg'

Cuvettes for aqueous solution samples, For use with 3M™ Clean-Trace™ Biomass Detection Kit

  • Gives real-time indication of total biological load allowing immediate corrective action
  • Ideal for process water, wastewater, water tanks and cooling towers
  • Offers flexibility when dealing with challenging samples

Specifications

Format Kit
Product Type Detection Cuvette
Test Type Environmental Test
For Use With (Application) Holds aqueous sample solutions such as water, wastewater, water tanks, cooling tanks
For Use With (Equipment) 3M™ Clean-Trace™ Biomass Detection Kit
Quantity 1000/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.