Learn More
Zymo Research Corporation Sample ZymoBIOMICS™ RNA Mini Kit (5 Preps)
SDP
Supplier:  Zymo Research Corporation R2001S
Encompass
Free Sample The ZymoBIOMICS RNA Miniprep Kit is designed for purifying RNA from a wide array of sample inputs that is ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae). The procedure uses Zymo-Spin Column technology that results in high-quality total RNA (including small RNAs 17-200 nt) that is free of PCR inhibitors and is ready for RT-PCR, hybridization, sequencing, etc.DNase I included.
Catalog No. NC3345988
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.