Learn More
Research Products International Corp BioMixer 3D Shaker w Dmpld Mat
SDP
Supplier:  Research Products International Corp 248590
Encompass_Preferred
The BioMixer combines the motions of an orbital and rocking shaker to produce a three dimensional and "nutating" gyration. Provides a gentle, thorough action without causing foaming. Ideal for re-suspending fragile DNA fragments and mixing of biological samples. Even the smallest single platform configuration has 24% more platform work surface than traditional nutating mixers. These mixers feature maintenance free, brushless motors capable of loads up to 2 kilograms and are safe for use in cold rooms and incubators.
Catalog No. 50-238-8431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.