Learn More
Biodesign Inc of Ny BioPrepNylon, for coarse initial filtration of biological preparations, 50 micron pore, pure nylon woven filter, roll 4 x 3 feet, reusable, autoclavable
Supplier:  Biodesign Inc of Ny N307
Encompass
BioPrepNylon is specifially designed for the coarse initial filtration of biological preparations. A pure nylon woven filter that is reusable and autoclavable with a 50 micron pore size. For most applications, BioPrepNylon is loosely placed in a Sieve Holder to form a funnel. The sample is placed in the funnel and allowed to drain. The layer of BioPrepNylon are then pressed together to remove all of the extraction liquid. As BioPrepNylon does not wet, significantly greater quantities of the extraction liquid are recovered compared to any other material. A pure nylon filter that does not bind proteins and without detergents, dyes, or salts that can be extracted.
Catalog No. NC2545809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.