Learn More
Ted Pella Inc Biopunch, ID 8.0mm, OD 8.50mm, 5/pk
SDP
Supplier:  Ted Pella Inc 1511180
Encompass
The Biopunch is a coring tool designed to core, punch and retrieve samples from thin or soft substrate source materials such as tissue, gels, paper, cloth, leaves, paint chips and films. The Biopunch consists of a sure grip hollow plastic embossed handle with a razor sharp seamless stainless steel cutting tip protected by a clear removable cap. Each Biopunch is individually packaged sterile with lot number and expiration date clearly imprinted on the package.
Catalog No. NC0669801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.