Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Biosate™ Peptone
GREENER_CHOICE

Catalog No. 294312
Change view
Click to view available options
Quantity:
454 g
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-431-2 25 lb.
B11862 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-431-2 Supplier Gibco™ Supplier No. 294312
Add to Cart
Add to Cart

An animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract.

The combination of pancreatic digest of casein and yeast extract provides synergistic nutritional benefits that are not provided by the components alone, resulting in a unique nutritional profile. Customers have shown the combined use of these two peptones, when supplied as Biosate Peptone, improves toxin production with Clostridial species for vaccines, such as Clostridium botulinum and Clostridium perfringens.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere
Product Line Biosate
Quantity 25 lb.
Source Animal Origin, casein and yeast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.