Learn More
Sartorius Biosensor / Amine Reactive 2nd Generation, 5/pk
Catalog No. NC1832858 Shop All Sartorius Products
Encompass
Amine Reactive Second Generation (AR2G) Biosensors - Trays of 96 biosensors compatible with amine reactive coupling chemistry for kinetic analysis. Requires amine coupling second generation kit.
Catalog No. NC1832858 Supplier Sartorius Supplier No. 185093
Amine Reactive Second Generation (AR2G) Biosensors - Trays of 96 biosensors compatible with amine reactive coupling chemistry for kinetic analysis. Requires amine coupling second generation kit.