Learn More
Sartorius Biosensor / Amine Reactive 2nd Generation (AR2G) Tray
Catalog No. NC0163309 Shop All Sartorius Products
Encompass
Amine Reactive 2nd Generation (AR2G) Biosensors - One tray of 96 biosensors for binding proteins compatible with amine reactive coupling chemistry. For kinetics applications. Requires amine coupling second generation kit (18-5095).
Catalog No. NC0163309 Supplier Sartorius Supplier No. 185092
Amine Reactive 2nd Generation (AR2G) Biosensors - One tray of 96 biosensors for binding proteins compatible with amine reactive coupling chemistry. For kinetics applications. Requires amine coupling second generation kit (18-5095).