Learn More
Sartorius BIOSENSOR/PROTEIN G PACK
Catalog No. NC9067688 Shop All Sartorius Products
Encompass
Protein G (ProG) Biosensors - Five trays of 96 biosensors coated with Protein G.
Catalog No. NC9067688 Supplier Sartorius Supplier No. 185083
Protein G (ProG) Biosensors - Five trays of 96 biosensors coated with Protein G.